Chlortetracyclin Hydrochlorid

Lieferant: LGC Standards PROMOCHEM
Gefahr

MM0607.00
PROOMM0607.00EA 1220 EUR
PROOMM0607.00
Chlortetracyclin Hydrochlorid
Chlortetracyclin Hydrochlorid
Formel: C₂₂H₂₃ClN₂O₈·HCl
Lagertemperatur: Kühlschrank
MDL: MFCD26142699
CAS-Nummer: 64-72-2
EINECS: 200-591-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR