Betain Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

MM0611.00
PROOMM0611.00EA 0 EUR
PROOMM0611.00
Betain Hydrochlorid
Betain Hydrochlorid
Formel: C₅H₁₂ClNO₂
Molecular Weight: 153,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011903
CAS-Nummer: 590-46-5
EINECS: 209-683-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR