2-(tert-Butylamino)-4'-chlorpropiophenon Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
MM0745.02
PROOMM0745.02EA
835
EUR
PROOMM0745.02
2-(tert-Butylamino)-4'-chlorpropiophenon Hydrochlorid
2-(tert-Butylamino)-4'-chlorpropiophenon Hydrochlorid
Formel:
C₁₃H₁₉Cl₂NO |
CAS-Nummer:
1049718-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...