2-(tert-Butylamino)-3'-brompropiophenon Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
MM0745.03
PROOMM0745.03EA
202
EUR
PROOMM0745.03
2-(tert-Butylamino)-3'-brompropiophenon Hydrochlorid
2-(tert-Butylamino)-3'-brompropiophenon Hydrochlorid
Formel:
C₁₃H₁₉BrClNO |
CAS-Nummer:
1049718-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...