2-(tert-Butylamino)propiophenon Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

MM0745.05
PROOMM0745.05EA 1810 EUR
PROOMM0745.05
2-(tert-Butylamino)propiophenon Hydrochlorid
2-(tert-Butylamino)propiophenon Hydrochlorid
Formel: C₁₃H₂₀ClNO
CAS-Nummer: 63199-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR