β-Chlorpropiophenon
Lieferant: LGC Standards PROMOCHEM
Synonyme:
3-Chlorpropiophenon
MM0745.08
PROOMM0745.08EA
99
EUR
PROOMM0745.08
β-Chlorpropiophenon
β-Chlorpropiophenon
Formel:
(C₆H₅)C(O)CH₂CH₂Cl Molecular Weight: 168,62 g/mol Siedepunkt: 263 °C (1013 hPa) Schmelzpunkt: 48…49 °C Dichte: 1,181 g/cm³ (20 °C) |
MDL:
MFCD00000990 CAS-Nummer: 936-59-4 EINECS: 213-317-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...