Ethosuximid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
2-Ethyl-2-methylsuccinimide, 3-Ethyl-3-methylpyrrolidine-2,5-dione
MM0750.00
PROOMM0750.00EA
248
EUR
PROOMM0750.00
Ethosuximid
Ethosuximid
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,17 g/mol |
MDL:
MFCD00072123 CAS-Nummer: 77-67-8 EINECS: 201-048-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...