5-Iod-2'-deoxyuridin (Idoxuridin)

Lieferant: LGC Standards PROMOCHEM

Synonyme: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

MM0791.00
PROOMM0791.00EA 261 EUR
PROOMM0791.00
5-Iod-2'-deoxyuridin (Idoxuridin)
5-Iod-2'-deoxyuridin (Idoxuridin)
Formel: C₉H₁₁IN₂O₅
Molecular Weight: 354,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134656
CAS-Nummer: 54-42-2
EINECS: 200-207-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR