(±)-3-Chinuclidinol
Lieferant: LGC Standards PROMOCHEM
Synonyme:
(±)-3-Hydroxy-1-azabicyclo[2.2.2]octan, (±)-3-Hydroxychinuclidin, (±)-Chinuclidin-3-ol
MM0825.02
PROOMM0825.02EA
1310
EUR
PROOMM0825.02
(±)-3-Chinuclidinol
(±)-3-Chinuclidinol
Formel:
C₇H₁₃NO Molecular Weight: 127,19 g/mol Schmelzpunkt: 225…227 °C |
MDL:
MFCD00151326 CAS-Nummer: 1619-34-7 EINECS: 216-578-4 UN: 3263 ADR: 8,III Merck Index: 12,08266 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...