2-Mercapto-1-methylimidazol

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-Mercapto-1-methyl-1H-imidazol, Thiamazol, 2-Mercapto-N-methyl-1H-imidazol, Methimazol, 2-Mercapto-N-methylimidazol

MM0889.00
PROOMM0889.00EA 1180 EUR
PROOMM0889.00
2-Mercapto-1-methylimidazol
2-Mercapto-1-methylimidazole
Formel: C₄H₆N₂S
Molecular Weight: 114,17 g/mol
Siedepunkt: 155 °C (1013 hPa)
Dichte: 1,36 g/cm³ (20 °C)
MDL: MFCD00179321
CAS-Nummer: 60-56-0
EINECS: 200-482-4
Merck Index: 13,06000

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR