1-(2,6-Dichlorphenyl)-2-thioharnstoff

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: 2,6-Dichlorphenylthioharnstoff, N-(2,6-Dichlorphenyl)thioharnstoff

MM0983.02
PROOMM0983.02EA 383 EUR
PROOMM0983.02
1-(2,6-Dichlorphenyl)-2-thioharnstoff
1-(2,6-Dichlorphenyl)-2-thioharnstoff
Formel: (C₆H₃Cl₂)NC(S)NH₂
Molecular Weight: 221,11 g/mol
Siedepunkt: 326 °C (1013 hPa)
Schmelzpunkt: 160…162 °C
MDL: MFCD00041156
CAS-Nummer: 6590-91-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR