1-Methyl-3-phenylpiperazin

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: MEFPIP

MM1005.07
PROOMM1005.07EA 1200 EUR
PROOMM1005.07
1-Methyl-3-phenylpiperazin
1-Methyl-3-phenylpiperazin
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
MDL: MFCD03411603
CAS-Nummer: 5271-27-2
EINECS: 431-180-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR