2,3-Diaminophenazin

Lieferant: LGC Standards PROMOCHEM

Synonyme: Phenazin-2,3-diyldiamin, DAP

MM2500.01
PROOMM2500.01EA 1830 EUR
PROOMM2500.01
2,3-Diaminophenazin
2,3-Diaminophenazine
Formel: C₁₂H₁₀N₄
Molecular Weight: 210,24 g/mol
MDL: MFCD00160693
CAS-Nummer: 655-86-7
EINECS: 211-512-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR