Meat homogenate - nutrients, NIST-1546A
Lieferant: LGC Standards PROMOCHEM
NIST-1546A
PROONIST-1546AEA 2150 EUR
PROONIST-1546A
Meat homogenate - nutrients, NIST-1546A
Standards Lebensmittel-Standards
This standard reference material is intended for determining fatty acids, cholestrol, proximates, calories, elements, vitamins and amino acids in canned meat products and similiar materials. The meat homogenate is a mixture of pork and chicken products blended together in a commercial process.
Standards: Traceable to NIST and SRM numbers.