Whole milk powder, NIST-1549A
Lieferant: LGC Standards PROMOCHEM
NIST-1549A
PROONIST-1549AEA 1970 EUR
PROONIST-1549A
Whole milk powder, NIST-1549A
Standards Lebensmittel-Standards
This reference standard is intended for validation of methods for determining proximates, fatty acids, cholestrol, vitamins, elements and amino acids in whole milk powder and similar materials.
Standards: Traceable to NIST and SRM numbers.