Residual fuel oil - sulphur, NIST-1622E reference material
Lieferant: LGC Standards PROMOCHEM
NIST-1622E
PROONIST-1622EEA 768 EUR
PROONIST-1622E
Residual fuel oil - sulphur, NIST-1622E reference material
Standards Mineralölindustrie-Standards
This reference standards are intended for use in the calibration of instruments and the evaluation of methods used in the determination of total sulphur in fuel oils and materials of similar matrix.
A unit consists of 100 ml of commercial No.6 residual fuel oil as defined by ASTM D396-95 standard specification for fuel oils.
Standards: Traceable to NIST and SRM numbers.