Coal fly ash - crace and constituent elements
Lieferant: LGC Standards PROMOCHEM
NIST-1633C
PROONIST-1633CEA 1260 EUR
PROONIST-1633C
Coal fly ash - crace and constituent elements
Standards Mineralölindustrie-Standards
This reference standard is intended for use in the evaluation of analytical methods for the determination of constituent elements in coal fly ash and materials of similar matrix. These are bituminous coal fly ash, sieved through a nominal sieve opening of 74 µm (200 mesh) and then blended to assure homogeneity.
A unit consists of 75 g of powdered material.
Standards: Traceable to NIST.