Low alloy steel standard
Lieferant: LGC Standards PROMOCHEM
NIST-1764A
PROONIST-1764AEA
1570
EUR
PROONIST-1764A
Low alloy steel standard
Standards
Standards, Legierungen und Metalle
This reference material is intended for use in the evaluation of chemical and instrumental analysis and in calibration of those methods.
- 22 analytes
A unit consists of a disk approximately 34 mm in diameter and 19 mm thick.
Standards: Traceable to NIST and SRM.