Lithium ore, spodumene - lithium oxide, NIST standards
Lieferant: LGC Standards PROMOCHEM
NIST-182
PROONIST-182EA
608
EUR
PROONIST-182
Lithium ore, spodumene - lithium oxide, NIST standards
Standards
Standards, Legierungen und Metalle
This reference standards are intended to use in checking the accuracy of assay methods. These are supplied in fine powder form.
Standards: Traceable to NIST and SRM numbers.