Siliciumnitrid

Lieferant: LGC Standards PROMOCHEM

Synonyme: Silicon(IV) nitride

NIST-1900
PROONIST-1900EA 666 EUR
PROONIST-1900
Siliciumnitrid
Siliciumnitrid
Formel: N₄Si₃
Molecular Weight: 140,28 g/mol
MDL: MFCD00011230
CAS-Nummer: 12033-89-5
EINECS: 234-796-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR