Peanut butter, NIST-2387 standard
Lieferant: LGC Standards PROMOCHEM
NIST-2387
PROONIST-2387EA
1580
EUR
PROONIST-2387
Peanut butter, NIST-2387 standard
Standards
Lebensmittel-Standards
This reference material intended to use in validating methods for determining fat, fatty acids, trace elements and tocopherol.
Standards: Traceable to NIST and SRM.