Standards for Lead paint films for childrens' products, NIST-2569
Lieferant: LGC Standards PROMOCHEM
NIST-2569
PROONIST-2569EA 1090 EUR
PROONIST-2569
Standards for Lead paint films for childrens' products, NIST-2569
Standards Umweltanalyse-Standards
Analyte details: lead at <0,2 ppm | 85,0 ppm | 314,4 ppm
Standards: Traceable to NIST and SRM.