Beryllium standard solution
Lieferant: LGC Standards PROMOCHEM
NIST-3105A
PROONIST-3105AEA 1070 EUR
PROONIST-3105A
Beryllium standard solution
Standards Kationischer Standards Beryllium-Standards
This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of beryllium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of beryllium.
Standards: Traceable to NIST and SRM numbers.