Cadmium standard solution, NIST-3108
Lieferant: LGC Standards PROMOCHEM
This reference standard is intended for use as a primary calibration standard for the quantitative determination of cadmium.
A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.
Standards: Traceable to NIST and SRM numbers.