Calcium standard solution
Lieferant: LGC Standards PROMOCHEM
NIST-3109A
PROONIST-3109AEA 1070 EUR
PROONIST-3109A
Calcium standard solution
Standards Kationischer Standards Calcium-Standards
This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of calcium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified solution prepared gravimetrically to contain a known mass fraction of calcium.
Standards: Traceable to NIST and SRM numbers.