Iron standard solution
Lieferant: LGC Standards PROMOCHEM
NIST-3126A
PROONIST-3126AEA 1170 EUR
PROONIST-3126A
Iron standard solution
Standards Kationischer Standards Eisen-Standards
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of iron.
A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.
Standards: Traceable to NIST and SRM numbers.