Nickel standard solutions
Lieferant: LGC Standards PROMOCHEM
NIST-3136
PROONIST-3136EA 1070 EUR
PROONIST-3136
Nickel standard solutions
Standards Kationischer Standards Nickel-Standards
This reference standard material is intended for use as a primary calibration standard for the quantitative determination of nickel.
A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of nickel.
Standards: Traceable to NIST and SRM numbers.