Potassium standard solution, NIST-3141A
Lieferant: LGC Standards PROMOCHEM
NIST-3141A
PROONIST-3141AEA 1410 EUR
PROONIST-3141A
Potassium standard solution, NIST-3141A
Standards Kationischer Standards Kalium-Standards
This reference standard is intended for use as a primary calibration standard for the quantitative determination of potassium.
A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.
Standards: Traceable to NIST and SRM numbers.