Sodium standard solution, NIST-3152A
Lieferant: LGC Standards PROMOCHEM
NIST-3152A
PROONIST-3152AEA 1070 EUR
PROONIST-3152A
Sodium standard solution, NIST-3152A
Standards Kationischer Standards Natrium-Standards
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of sodium.
A unit consists of 50 ml of an aqueous solution in a high-density polyethylene bottle sealed in an aluminised bag.
Standards: Traceable to NIST and SRM numbers.