Zinc standard solution
Lieferant: LGC Standards PROMOCHEM
NIST-3168A
PROONIST-3168AEA 1080 EUR
PROONIST-3168A
Zinc standard solution
Standards Kationischer Standards Zink-Standards
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of zinc.
A unit consists of 50 ml of a single-element solution in a high-density polyethylene bottle sealed in an aluminised bag.
Standards: Traceable to NIST and SRM numbers.