Glass - trace elements, NIST standards
Lieferant: LGC Standards PROMOCHEM
NIST-610
NIST-611
NIST-612
NIST-613
NIST-614
NIST-616
NIST-617
PROONIST-610EA
950
EUR
PROONIST-610
PROONIST-611
PROONIST-612
PROONIST-613
PROONIST-614
PROONIST-616
PROONIST-617
Glass - trace elements, NIST standards
Standards
Umweltanalyse-Standards
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Standards: Traceable to NIST and SRM.