Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Lieferant: LGC Standards PROMOCHEM
NIST-68C
PROONIST-68CEA
813
EUR
PROONIST-68C
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Standards
Standards, Legierungen und Metalle
This standard reference material is in the powder form intended for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis.
- 8 analytes/elements measured
Standards: Traceable to NIST and SRM.