Iron ore
Lieferant: LGC Standards PROMOCHEM
NIST-690
NIST-691
NIST-692
NIST-693
PROONIST-690EA
688
EUR
PROONIST-690
PROONIST-691
PROONIST-692
PROONIST-693
Iron ore
Standards
Kationischer Standards
Eisen-Standards
This reference standard is intended for use in checking chemical methods of analysis and in the calibration with instrumental methods of analysis.
Standards: Traceable to NIST.