Organic reference standards, HMMNI (nitroimidazole metabolite NMIA P1296)
Lieferant: LGC Standards PROMOCHEM
NMIAP1296
PROONMIAP1296EA
445
EUR
PROONMIAP1296
Organic reference standards, HMMNI (nitroimidazole metabolite NMIA P1296)
Standards
Organische Standards
Purity estimate obtained is 99,8 ± 1,3% mass fraction, (95% coverage interval).
- Pale yellow crystals
- Moisture content ≤0,1% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,2% mass fraction
- CAS no.: 936-05-0
Synonyms: 2-hydroxymethyl-1-methyl-5-nitro-imidazole; 1-methyl-5-nitro-imidazole-2-methanol; 1-methyl-5-nitro-2-imidazole methanol
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...