cis-Chlordan

Lieferant: LGC Standards PROMOCHEM

NMIAP1624
PROONMIAP1624EA 303 EUR
PROONMIAP1624
cis-Chlordan
cis-Chlordane
Formel: C₁₀H₆Cl₈
CAS-Nummer: 5103-71-9
EINECS: 225-825-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR