trans-Chlordan
Lieferant: LGC Standards PROMOCHEM
NMIAP1625
PROONMIAP1625EA
303
EUR
PROONMIAP1625
trans-Chlordan
trans-Chlordane
Formel:
C₁₀H₆Cl₈ |
CAS-Nummer:
5103-74-2 EINECS: 225-826-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...