Organic reference standards, fipronil sulphone (NMIA P1731)
Lieferant: LGC Standards PROMOCHEM
NMIAP1731
PROONMIAP1731EA
441
EUR
PROONMIAP1731
Organic reference standards, fipronil sulphone (NMIA P1731)
Standards
Organische Standards
Purity estimate obtained from a combination of traditional analytical techniques is 99,3 ± 0,6% mass fraction, (95% coverage interval). The purity estimate by traditional analytical techniques was obtained by subtraction from 100% of total impurities by GC-FID, and Karl Fischer analysis. Supporting evidence is provided by elemental microanalysis and 1H NMR analysis.
- White solid
- Moisture content ≤0,2% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,2% mass fraction
Synonym: 5-amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-4-((trifluoromethyl)sulphonyl)pyrazole-3-carbonitrile.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...