Organic reference standards, α-zearalenol (NMIA P1795)

Lieferant: LGC Standards PROMOCHEM

NMIAP1795
PROONMIAP1795EA 451 EUR
PROONMIAP1795
Organic reference standards, α-zearalenol (NMIA P1795)
Standards Organische Standards

Purity estimate obtained by subtraction from 100% of total impurities by GC-FID, thermogravimetric analysis, Karl Fischer analysis and 1H NMR is 99,1 ± 1,4% mass fraction, (95% coverage interval). Supporting evidence is provided by elemental microanalysis.


  • White crystalline powder
  • Moisture content ≤0,5% mass fraction
  • Volatile residue ≤0,1% mass fraction
  • Non volatile residue ≤0,2% mass fraction
  • CAS no.: 36455-72-8


Synonyms: (3S,7R,11E)-3,4,5,6,7,8,9,10-octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR