Laktose
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Milchzucker, 4-(β-D-Galactopyranosyl)-D-glucopyranose
RTC-PHR-1025
PROORTC-PHR-1025EA
0
EUR
PROORTC-PHR-1025
Laktose
Laktose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Siedepunkt: 592 Schmelzpunkt: 222,8 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151251 CAS-Nummer: 63-42-3 EINECS: 200-559-2 Merck Index: 12,05356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...