Acetaldehyd 95
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Ethanal
TRC-A132600-1G
PROOTRC-A132600-1GEA
199
EUR
PROOTRC-A132600-1G
Acetaldehyd 95
Acetaldehyd
Formel:
CH₃CHO Molecular Weight: 44.05 g/mol Siedepunkt: 20.1 Schmelzpunkt: -123 Flammpunkt: -27 |
MDL:
MFCD00006991 CAS-Nummer: 75-07-0 Merck Index: 12,00037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...