L-(+)-Gulonic acid-γ-lactone

Lieferant: LGC Standards PROMOCHEM

Synonyme: L-Gulonic acid-1,4-lactone, (3S,4R,5R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one, L-Gulonolactone

TRC-G855000-5G
PROOTRC-G855000-5GEA 97 EUR
PROOTRC-G855000-5G
L-(+)-Gulonic acid-γ-lactone
L-(+)-Gulonic acid-gamma-lactone
Formel: C₆H₁₀O₆
Molecular Weight: 178.14 g/mol
Schmelzpunkt: 184…186 °C
Lagertemperatur: Room Temperature
MDL: MFCD00064331
CAS-Nummer: 1128-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR