Alternariol monomethyl ether
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
TRCA575770-5MG
PROOTRCA575770-5MGEA
672
EUR
PROOTRCA575770-5MG
Alternariol monomethyl ether
Alternariol monomethyl ether
Formel:
C₁₅H₁₂O₅ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058532 CAS-Nummer: 23452-05-3 EINECS: 636-657-0 UN: 3462 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...