Meloxicam
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
TRCM216100-250
PROOTRCM216100-250EA
0
EUR
PROOTRCM216100-250
Meloxicam
Meloxicam
Formel:
C₁₄H₁₃N₃O₄S₂ Molecular Weight: 351,40 g/mol |
MDL:
MFCD00868752 CAS-Nummer: 71125-38-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Pale yellow to light yellow solid |
NMR | Conforms |
Elemental analysis | %C: 47.69, %H: 3.58, %N: 11.73 |
HPLC purity | 99.88% (355 nm) |
MS | Conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...