Nicardipin -hydrochlorid, gelb, fest

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, Nicardipine HCl

TRCN394500-50
PROOTRCN394500-50EA 63 EUR
PROOTRCN394500-50
Nicardipin -hydrochlorid, gelb, fest
Nicardipin -hydrochlorid
Formel: C₂₆H₂₉N₃O₆
Molecular Weight: 515,99 g/mol
Schmelzpunkt: 176,..178 °C
MDL: MFCD00057327
CAS-Nummer: 54527-84-3
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Crystalline powder Appearance
Solubility DMSO (Slightly), Methanol (Slightly)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR