Oxymetazoline hydrochloride, weiß
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
SOxymetazoline hydrochloride, 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrin hydrochloride
TRCO876470-100
PROOTRCO876470-100EA
0
EUR
PROOTRCO876470-100
Oxymetazoline hydrochloride, weiß
Oxymetazoline hydrochloride
Formel:
C₁₆H₂₄N₂O·HCl Molecular Weight: 296,839 g/mol Schmelzpunkt: >270 °C |
MDL:
MFCD00058147 CAS-Nummer: 2315-02-8 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...