Pyridoxin Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

Synonyme: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

TRCP991735-1G
PROOTRCP991735-1GEA 99.1 EUR
PROOTRCP991735-1G
Pyridoxin Hydrochlorid
Pyridoxin Hydrochlorid
Formel: C₈H₁₂ClNO₃
Molecular Weight: 205,64 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012807
CAS-Nummer: 58-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR