Reference materials, cloud point reference, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-C5-250
PROOVHG-C5-250EA
301
EUR
PROOVHG-C5-250
Reference materials, cloud point reference, VHG
Standards
Mineralölindustrie-Standards
These reference materials are intended for use in accordance with ASTM D2500 for the determination of the cloud point of petroleum products. The cloud point of a petroleum product is an index of the lowest temperature of their utility for certain applications.
Lieferumfang: Each standard is supplied with a full certificate of analysis (COA) that states the certified value.
Bitte beachten Not intended for use with phase instruments.