XRF thin film, Kapton®, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-FKP30-R15
PROOVHG-FKP30-R15EA 172 EUR
PROOVHG-FKP30-R15
XRF thin film, Kapton®, VHG
Versiegeln Verschlussfolien
XRF thin film of 7,6 µm thickness and 76,2×15240 mm roll.