Chlorite ion chromatography standards, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-ICLO2-100 VHG-ICLO2-500
PROOVHG-ICLO2-100EA 58 EUR
PROOVHG-ICLO2-100 PROOVHG-ICLO2-500
Chlorite ion chromatography standards, CRM, VHG
Standards Anionische Standards Chlorit-Standards
This solution is intended for use as a certified reference material or calibration standard, for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST.