Formate ion chromatography standards, VHG
Lieferant: LGC Standards PROMOCHEM
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and CRM.